Drukuj
Anti-ALIX Rabbit Monoclonal Antibody [clone: EPR23653-32]
Dostawca: Abcam
AB275377-100 AB275377-1ML
ABCAAB275377-100EA 3200 PLN
ABCAAB275377-100 ABCAAB275377-1ML
Anti-ALIX Rabbit Monoclonal Antibody [clone: EPR23653-32]
Przeciwciała
Rabbit monoclonal [EPR23653-32] to ALIX.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ALIX
Clonality: Monoclonal
Clone: EPR23653-32
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat