Drukuj
Anti-CD63 Rabbit Monoclonal Antibody [clone: EPR22458-280]
Dostawca: Abcam
AB254011-100 AB254011-1
ABCAAB254011-100EA 4530 PLN
ABCAAB254011-100 ABCAAB254011-1
Anti-CD63 Rabbit Monoclonal Antibody [clone: EPR22458-280]
Przeciwciała
Rabbit monoclonal [EPR22458-280] to CD63 - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: EPR22458-280
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human