Drukuj

Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR24411-22]

Dostawca: Abcam
AB270972-100 AB270972-20UL AB270972-1ML
ABCAAB270972-100EA 3170 PLN
ABCAAB270972-100 ABCAAB270972-20UL ABCAAB270972-1ML
Anti-AIRE Rabbit Monoclonal Antibody [clone: EPR24411-22]
Przeciwciała
Rabbit monoclonal [EPR24411-22] to AIRE.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AIRE
Clonality: Monoclonal
Clone: EPR24411-22
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR