Drukuj

Anti-EpCAM Mouse Monoclonal Antibody [clone: AUA1]

Dostawca: Abcam
AB308057-10UG AB308057-100UG AB308057-20UG AB308057-1MG
ABCAAB308057-10UGEA 1030 PLN
ABCAAB308057-10UG ABCAAB308057-100UG ABCAAB308057-20UG ABCAAB308057-1MG
Anti-EpCAM Mouse Monoclonal Antibody [clone: AUA1]
Przeciwciała
Mouse monoclonal [AUA1] to EpCAM.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: AUA1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR