Drukuj
Anti-KIF3A Rabbit Monoclonal Antibody [clone: EPR5087]
Dostawca: Abcam
AB248566-1 AB248566-100
ABCAAB248566-1EA 40660 PLN
ABCAAB248566-1 ABCAAB248566-100
Anti-KIF3A Rabbit Monoclonal Antibody [clone: EPR5087]
Przeciwciała
Rabbit monoclonal [EPR5087] to KIF3A - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: KIF3A
Clonality: Monoclonal
Clone: EPR5087
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human