Drukuj
Anti-DCTN1/p150-glued Mouse Monoclonal Antibody [clone: 12/P150GLUED]
Dostawca: Abcam
AB302965-1 AB302965-100
ABCAAB302965-1EA 40660 PLN
ABCAAB302965-1 ABCAAB302965-100
Anti-DCTN1/p150-glued Mouse Monoclonal Antibody [clone: 12/P150GLUED]
Przeciwciała
Mouse monoclonal [12/P150GLUED] to DCTN1/p150-glued.
  • BSA and Azide free
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DCTN1/p150-glued
Clonality: Monoclonal
Clone: 12/P150GLUED
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat