Drukuj

Anti-CD13 Mouse Monoclonal Antibody [clone: WM15]

Dostawca: Abcam
AB252261-100 AB252261-1
ABCAAB252261-100EA 4530 PLN
ABCAAB252261-100 ABCAAB252261-1
Anti-CD13 Mouse Monoclonal Antibody [clone: WM15]
Przeciwciała
Mouse monoclonal [WM15] to CD13 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: WM15
Conjugation:
Epitope: The antibody recognizes an extracellular epitope.
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR