Drukuj

Anti-ST2 Mouse Monoclonal Antibody [clone: B-L42]

Dostawca: Abcam
AB288543-200 AB288543-500
ABCAAB288543-200EA 3500 PLN
ABCAAB288543-200 ABCAAB288543-500
Anti-ST2 Mouse Monoclonal Antibody [clone: B-L42]
Przeciwciała
Mouse monoclonal [B-L42] to ST2 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ST2
Clonality: Monoclonal
Clone: B-L42
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR