Drukuj

Anti-PD1 Rabbit Recombinant Multiclonal Antibody [clone: RM1033]

Dostawca: Abcam
AB309363-100UL AB309363-20UL AB309363-1ML
ABCAAB309363-100ULEA 2590 PLN
ABCAAB309363-100UL ABCAAB309363-20UL ABCAAB309363-1ML
Anti-PD1 Rabbit Recombinant Multiclonal Antibody [clone: RM1033]
Przeciwciała
Rabbit recombinant multiclonal [RM1033] to PD1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PD1
Clonality: Recombinant Multiclonal
Clone: RM1033
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR