Drukuj

Anti-Proteasome 20S beta 6 Rabbit Monoclonal Antibody [clone: EPR9685]

Dostawca: Abcam
AB248944-1 AB248944-100
ABCAAB248944-1EA 40660 PLN
ABCAAB248944-1 ABCAAB248944-100
Anti-Proteasome 20S beta 6 Rabbit Monoclonal Antibody [clone: EPR9685]
Przeciwciała
Rabbit monoclonal [EPR9685] to Proteasome 20S beta 6 - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Proteasome 20S beta 6
Clonality: Monoclonal
Clone: EPR9685
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR