Drukuj

Anti-Collagen VI Rabbit Monoclonal Antibody [clone: EPR17072]

Dostawca: Abcam
AB271938-100 AB271938-1
ABCAAB271938-100EA 4530 PLN
ABCAAB271938-100 ABCAAB271938-1
Anti-Collagen VI Rabbit Monoclonal Antibody [clone: EPR17072]
Przeciwciała
Rabbit monoclonal [EPR17072] to Collagen VI - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Collagen VI
Clonality: Monoclonal
Clone: EPR17072
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR