Drukuj

Anti-MPG/AAG Rabbit Monoclonal Antibody [clone: EPR10959(B)]

Dostawca: Abcam
AB155092-100 AB155092-10 AB155092-1ML
ABCAAB155092-100EA 3670 PLN
ABCAAB155092-100 ABCAAB155092-10 ABCAAB155092-1ML
Anti-MPG/AAG Rabbit Monoclonal Antibody [clone: EPR10959(B)]
Przeciwciała
Rabbit monoclonal [EPR10959(B)] to MPG/AAG.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MPG/AAG
Clonality: Monoclonal
Clone: EPR10959(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR