Drukuj
Anti-Sortilin/NT3 Rabbit Monoclonal Antibody [clone: EPR23093-58]
Dostawca: Abcam
AB263873-100 AB263873-1
ABCAAB263873-100EA 4530 PLN
ABCAAB263873-100 ABCAAB263873-1
Anti-Sortilin/NT3 Rabbit Monoclonal Antibody [clone: EPR23093-58]
Przeciwciała
Rabbit monoclonal [EPR23093-58] to Sortilin/NT3 - BSA and Azide free.
Type: Primary
Antigen: Sortilin/NT3
Clonality: Monoclonal
Clone: EPR23093-58
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat