Drukuj
Anti-Glycerol 3 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR25996-4]
Dostawca: Abcam
AB313871-100UL AB313871-20UL AB313871-1ML
ABCAAB313871-100ULEA 3090 PLN
ABCAAB313871-100UL ABCAAB313871-20UL ABCAAB313871-1ML
Anti-Glycerol 3 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR25996-4]
Przeciwciała
Rabbit monoclonal [EPR25996-4] to Glycerol 3 Phosphate Dehydrogenase.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Glycerol 3 Phosphate Dehydrogenase
Clonality: Monoclonal
Clone: EPR25996-4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat