Drukuj

Anti-gamma H2A.X (phospho S139) Rabbit Monoclonal Antibody [clone: EP854(2)Y]

Dostawca: Abcam
AB215967-100 AB215967-1
ABCAAB215967-100EA 4530 PLN
ABCAAB215967-100 ABCAAB215967-1
Anti-gamma H2A.X (phospho S139) Rabbit Monoclonal Antibody [clone: EP854(2)Y]
Przeciwciała
Rabbit monoclonal [EP854(2)Y] to gamma H2A.X (phospho S139) - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: gamma H2A.X
Clonality: Monoclonal
Clone: EP854(2)Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR