Drukuj
Anti-NKG2D Rabbit Monoclonal Antibody [clone: EPR22935-131] (PE (Phycoerythrin))
Dostawca: Abcam
AB314290-100UL AB314290-10UL
ABCAAB314290-100ULEA 3640 PLN
ABCAAB314290-100UL ABCAAB314290-10UL
Anti-NKG2D Rabbit Monoclonal Antibody [clone: EPR22935-131] (PE (Phycoerythrin))
Przeciwciała
PE Rabbit monoclonal [EPR22935-131] to NKG2D.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: EPR22935-131
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human