Drukuj

Anti-L1CAM Rabbit Monoclonal Antibody [clone: EPR18750]

Dostawca: Abcam
AB208155-100 AB208155-10 AB208155-1ML
ABCAAB208155-100EA 3640 PLN
ABCAAB208155-100 ABCAAB208155-10 ABCAAB208155-1ML
Anti-L1CAM Rabbit Monoclonal Antibody [clone: EPR18750]
Przeciwciała
Rabbit monoclonal [EPR18750] to L1CAM.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: EPR18750
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR