Drukuj
Anti-NF-kB p65 (phospho S536) Rabbit Monoclonal Antibody [clone: NFKBp65S536-B7] (PE (Phycoerythrin))
Dostawca: Abcam
AB278630-10 AB278630-100
ABCAAB278630-10EA 1200 PLN
ABCAAB278630-10 ABCAAB278630-100
Anti-NF-kB p65 (phospho S536) Rabbit Monoclonal Antibody [clone: NFKBp65S536-B7] (PE (Phycoerythrin))
Przeciwciała
PE Rabbit monoclonal [NFKBp65S536-B7] to NF-kB p65 (phospho S536).
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: NF-kB p65
Clonality: Monoclonal
Clone: NFKBp65S536-B7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human