Drukuj

Anti-NDUFB11 Rabbit Monoclonal Antibody [clone: EPR15252]

Dostawca: Abcam
AB183716-100 AB183716-1ML
ABCAAB183716-100EA 2750 PLN
ABCAAB183716-100 ABCAAB183716-1ML
Anti-NDUFB11 Rabbit Monoclonal Antibody [clone: EPR15252]
Przeciwciała
Rabbit monoclonal [EPR15252] to NDUFB11.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NDUFB11
Clonality: Monoclonal
Clone: EPR15252
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR