Drukuj

Anti-Alpha-synuclein Mouse Monoclonal Antibody [clone: MJFR1]

Dostawca: Abcam
AB309158-1MG AB309158-100UG
ABCAAB309158-1MGEA 20070 PLN
ABCAAB309158-1MG ABCAAB309158-100UG
Anti-Alpha-synuclein Mouse Monoclonal Antibody [clone: MJFR1]
Przeciwciała
Mouse monoclonal [MJFR1] to Alpha-synuclein - Chimeric – BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Alpha-synuclein
Clonality: Monoclonal
Clone: MJFR1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR