Drukuj

Anti-Glucose Transporter GLUT4 Rabbit Monoclonal Antibody [clone: EPR26571-45]

Dostawca: Abcam
AB313775-100UL AB313775-20UL AB313775-1ML
ABCAAB313775-100ULEA 3090 PLN
ABCAAB313775-100UL ABCAAB313775-20UL ABCAAB313775-1ML
Anti-Glucose Transporter GLUT4 Rabbit Monoclonal Antibody [clone: EPR26571-45]
Przeciwciała
Rabbit monoclonal [EPR26571-45] to Glucose Transporter GLUT4.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glucose Transporter GLUT4
Clonality: Monoclonal
Clone: EPR26571-45
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR