Drukuj
Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR22311-17]
Dostawca: Abcam
AB242124-100 AB242124-20UL AB242124-1ML
ABCAAB242124-100EA 3470 PLN
ABCAAB242124-100 ABCAAB242124-20UL ABCAAB242124-1ML
Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR22311-17]
Przeciwciała
Rabbit monoclonal [EPR22311-17] to CD28.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: EPR22311-17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse