Drukuj

Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K20/78]

Dostawca: Abcam
AB252537-100 AB252537-1MG
ABCAAB252537-100EA 3390 PLN
ABCAAB252537-100 ABCAAB252537-1MG
Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K20/78]
Mouse monoclonal [K20/78] to Kv1.1 potassium channel.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Kv1.1 potassium channel
Clonality: Monoclonal
Clone: K20/78
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR