Drukuj

Anti-EGR2 Rabbit Monoclonal Antibody [clone: EPR23228-40]

Dostawca: Abcam
AB245228-100 AB245228-1ML
ABCAAB245228-100EA 3170 PLN
ABCAAB245228-100 ABCAAB245228-1ML
Anti-EGR2 Rabbit Monoclonal Antibody [clone: EPR23228-40]
Przeciwciała
Rabbit monoclonal [EPR23228-40] to EGR2.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EGR2
Clonality: Monoclonal
Clone: EPR23228-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR