Drukuj

Anti-SARS-CoV-2 Spike Glycoprotein S1 Mouse Monoclonal Antibody [clone: 8B12-C2]

Dostawca: Abcam
AB282749-100 AB282749-1
ABCAAB282749-100EA 4530 PLN
ABCAAB282749-100 ABCAAB282749-1
Anti-SARS-CoV-2 Spike Glycoprotein S1 Mouse Monoclonal Antibody [clone: 8B12-C2]
Przeciwciała
Mouse monoclonal [8B12-C2] to SARS-CoV-2 Spike Glycoprotein S1 - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 Spike Glycoprotein S1
Clonality: Monoclonal
Clone: 8B12-C2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR