Drukuj

Anti-LAS1L Rabbit Monoclonal Antibody [clone: EPR8988(B)]

Dostawca: Abcam
AB140656-100 AB140656-1ML
ABCAAB140656-100EA 3170 PLN
ABCAAB140656-100 ABCAAB140656-1ML
Anti-LAS1L Rabbit Monoclonal Antibody [clone: EPR8988(B)]
Przeciwciała
Rabbit monoclonal [EPR8988(B)] to LAS1L.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LAS1L
Clonality: Monoclonal
Clone: EPR8988(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR