Drukuj
Anti-Bcl 7A Mouse Monoclonal Antibody [clone: 15C]
Dostawca: Abcam
AB252800-1 AB252800-100
ABCAAB252800-1EA 40660 PLN
ABCAAB252800-1 ABCAAB252800-100
Anti-Bcl 7A Mouse Monoclonal Antibody [clone: 15C]
Przeciwciała
Mouse monoclonal [15C] to Bcl 7A - BSA and Azide free.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Bcl 7A
Clonality: Monoclonal
Clone: 15C
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human