Drukuj

Anti-PAX5 Rabbit Monoclonal Antibody [clone: SP34]

Dostawca: Abcam
AB227635-500 AB227635-100 AB227635-1
ABCAAB227635-500EA 4470 PLN
ABCAAB227635-500 ABCAAB227635-100 ABCAAB227635-1
Anti-PAX5 Rabbit Monoclonal Antibody [clone: SP34]
Przeciwciała
Rabbit monoclonal [SP34] to PAX5 - ChIP Grade.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PAX5
Clonality: Monoclonal
Clone: SP34
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR