Drukuj

Anti-CD22 Rabbit Monoclonal Antibody [clone: SP104]

Dostawca: Abcam
AB227645-1 AB227645-500 AB227645-100
ABCAAB227645-1EA 4780 PLN
ABCAAB227645-1 ABCAAB227645-500 ABCAAB227645-100
Anti-CD22 Rabbit Monoclonal Antibody [clone: SP104]
Przeciwciała
Rabbit monoclonal [SP104] to CD22 - C-terminal.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: SP104
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR