Drukuj

Anti-Rsk 2/MAPKAP Kinase 1b (phospho S227) Rabbit Monoclonal Antibody [clone: EPR2847Y]

Dostawca: Abcam
AB247405-1 AB247405-100
ABCAAB247405-1EA 40660 PLN
ABCAAB247405-1 ABCAAB247405-100
Anti-Rsk 2/MAPKAP Kinase 1b (phospho S227) Rabbit Monoclonal Antibody [clone: EPR2847Y]
Przeciwciała
Rabbit monoclonal [EPR2847Y] to Rsk 2 / MAPKAP Kinase 1b (phospho S227) - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Rsk 2 / MAPKAP Kinase 1b
Clonality: Monoclonal
Clone: EPR2847Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR