Drukuj

Anti-HP1 gamma/CBX3 Rabbit Monoclonal Antibody [clone: EPR10465(B)]

Dostawca: Abcam
AB249170-1 AB249170-100
ABCAAB249170-1EA 40660 PLN
ABCAAB249170-1 ABCAAB249170-100
Anti-HP1 gamma/CBX3 Rabbit Monoclonal Antibody [clone: EPR10465(B)]
Przeciwciała
Rabbit monoclonal [EPR10465(B)] to HP1 gamma/CBX3 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HP1 gamma/CBX3
Clonality: Monoclonal
Clone: EPR10465(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR