Drukuj

Anti-CD94 Rat Monoclonal Antibody [clone: 18D3]

Dostawca: Abcam
AB282717-100 AB282717-1
ABCAAB282717-100EA 4530 PLN
ABCAAB282717-100 ABCAAB282717-1
Anti-CD94 Rat Monoclonal Antibody [clone: 18D3]
Przeciwciała
Rat monoclonal [18D3] to CD94 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD94
Clonality: Monoclonal
Clone: 18D3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR