Drukuj
Anti-Collagen VI alpha 2 Rabbit Monoclonal Antibody [clone: EPR7889(N)]
Dostawca: Abcam
AB180855-100 AB180855-1ML
ABCAAB180855-100EA 3170 PLN
ABCAAB180855-100 ABCAAB180855-1ML
Anti-Collagen VI alpha 2 Rabbit Monoclonal Antibody [clone: EPR7889(N)]
Przeciwciała
Rabbit monoclonal [EPR7889(N)] to Collagen VI alpha 2.
Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Collagen VI alpha 2
Clonality: Monoclonal
Clone: EPR7889(N)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human