Drukuj

Anti-TRAP220/MED1 (phospho T1457) Rabbit Monoclonal Antibody [clone: EPR13269]

Dostawca: Abcam
AB250350-100 AB250350-1
ABCAAB250350-100EA 4530 PLN
ABCAAB250350-100 ABCAAB250350-1
Anti-TRAP220/MED1 (phospho T1457) Rabbit Monoclonal Antibody [clone: EPR13269]
Przeciwciała
Rabbit monoclonal [EPR13269] to TRAP220/MED1 (phospho T1457) - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRAP220/MED1
Clonality: Monoclonal
Clone: EPR13269
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR