Drukuj

Anti-Interferon regulatory factor 9/IRF-9 Mouse Monoclonal Antibody [clone: 6F1-H5]

Dostawca: Abcam
AB284665-1 AB284665-100
ABCAAB284665-1EA 40660 PLN
ABCAAB284665-1 ABCAAB284665-100
Anti-Interferon regulatory factor 9/IRF-9 Mouse Monoclonal Antibody [clone: 6F1-H5]
Przeciwciała
Mouse monoclonal [6F1-H5] to Interferon regulatory factor 9/IRF-9 - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Interferon regulatory factor 9/IRF-9
Clonality: Monoclonal
Clone: 6F1-H5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR