Drukuj

Anti-Cyclin B1 Rabbit Monoclonal Antibody [clone: EPR17060]

Dostawca: Abcam
AB181593-100 AB181593-1ML
ABCAAB181593-100EA 3500 PLN
ABCAAB181593-100 ABCAAB181593-1ML
Anti-Cyclin B1 Rabbit Monoclonal Antibody [clone: EPR17060]
Przeciwciała
Rabbit monoclonal [EPR17060] to Cyclin B1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cyclin B1
Clonality: Monoclonal
Clone: EPR17060
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR