Drukuj
Anti-CaMKII delta Rabbit Monoclonal Antibody [clone: EPR13095]
Dostawca: Abcam
AB181052-100 AB181052-1ML
ABCAAB181052-100EA 3470 PLN
ABCAAB181052-100 ABCAAB181052-1ML
Anti-CaMKII delta Rabbit Monoclonal Antibody [clone: EPR13095]
Przeciwciała
Rabbit monoclonal [EPR13095] to CaMKII delta.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CaMKII delta
Clonality: Monoclonal
Clone: EPR13095
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat