Drukuj

Anti-Polyoma virus, Large T antigen Mouse Monoclonal Antibody [clone: CM2B4]

Dostawca: Abcam
AB307450-100UG AB307450-20UG AB307450-1MG
ABCAAB307450-100UGEA 3090 PLN
ABCAAB307450-100UG ABCAAB307450-20UG ABCAAB307450-1MG
Anti-Polyoma virus, Large T antigen Mouse Monoclonal Antibody [clone: CM2B4]
Przeciwciała
Mouse monoclonal [CM2B4] to Polyoma virus, Large T antigen.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polyoma virus, Large T antigen
Clonality: Monoclonal
Clone: CM2B4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR