Drukuj

Anti-Inhibin alpha Rabbit Monoclonal Antibody [clone: EPY2782]

Dostawca: Abcam
AB226844-100 AB226844-1
ABCAAB226844-100EA 4530 PLN
ABCAAB226844-100 ABCAAB226844-1
Anti-Inhibin alpha Rabbit Monoclonal Antibody [clone: EPY2782]
Przeciwciała
Rabbit monoclonal [EPY2782] to Inhibin alpha - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Inhibin alpha
Clonality: Monoclonal
Clone: EPY2782
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR