Drukuj

Anti-BRAF Rabbit Monoclonal Antibody [clone: EP152Y]

Dostawca: Abcam
AB189351-100 AB189351-1
ABCAAB189351-100EA 4530 PLN
ABCAAB189351-100 ABCAAB189351-1
Anti-BRAF Rabbit Monoclonal Antibody [clone: EP152Y]
Przeciwciała
Rabbit monoclonal [EP152Y] to BRAF - BSA and Azide free.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: EP152Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR