Drukuj

Anti-PRDM1/Blimp1 Mouse Monoclonal Antibody [clone: ROS195G]

Dostawca: Abcam
AB252785-1 AB252785-100
ABCAAB252785-1EA 44210 PLN
ABCAAB252785-1 ABCAAB252785-100
Anti-PRDM1/Blimp1 Mouse Monoclonal Antibody [clone: ROS195G]
Przeciwciała
Mouse monoclonal [ROS195G] to PRDM1/Blimp1 - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PRDM1/Blimp1
Clonality: Monoclonal
Clone: ROS195G
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR