Drukuj

Anti-CD134/OX40L receptor Rabbit Monoclonal Antibody [clone: EPR17Y]

Dostawca: Abcam
AB244988-1 AB244988-100
ABCAAB244988-1EA 41350 PLN
ABCAAB244988-1 ABCAAB244988-100
Anti-CD134/OX40L receptor Rabbit Monoclonal Antibody [clone: EPR17Y]
Przeciwciała
Rabbit monoclonal [EPR17Y] to CD134/OX40L receptor - BSA and Azide free (Detector).

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD134/OX40L receptor
Clonality: Monoclonal
Clone: EPR17Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR