Drukuj

Anti-mCherry Rabbit Monoclonal Antibody [clone: EPR20579]

Dostawca: Abcam
AB213511-100 AB213511-1ML
ABCAAB213511-100EA 2110 PLN
ABCAAB213511-100 ABCAAB213511-1ML
Anti-mCherry Rabbit Monoclonal Antibody [clone: EPR20579]
Przeciwciała
Rabbit monoclonal [EPR20579] to mCherry.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: mCherry
Clonality: Monoclonal
Clone: EPR20579
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR