Drukuj

Anti-Cenexin1/ODF2 Rabbit Monoclonal Antibody [clone: EPR13561]

Dostawca: Abcam
AB186412-100 AB186412-1ML
ABCAAB186412-100EA 3250 PLN
ABCAAB186412-100 ABCAAB186412-1ML
Anti-Cenexin1/ODF2 Rabbit Monoclonal Antibody [clone: EPR13561]
Przeciwciała
Rabbit monoclonal [EPR13561] to Cenexin1/ODF2.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cenexin1/ODF2
Clonality: Monoclonal
Clone: EPR13561
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR