Drukuj

Anti-PD1 Rat Monoclonal Antibody [clone: 29F.1A12]

Dostawca: Abcam
AB309360-100UG AB309360-20UG AB309360-1MG
ABCAAB309360-100UGEA 3090 PLN
ABCAAB309360-100UG ABCAAB309360-20UG ABCAAB309360-1MG
Anti-PD1 Rat Monoclonal Antibody [clone: 29F.1A12]
Przeciwciała
Rat monoclonal [29F.1A12] to PD1.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: 29F.1A12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR