Drukuj

Anti-Vitamin D Binding protein Rabbit Monoclonal Antibody [clone: EP2923Y]

Dostawca: Abcam
AB247521-1 AB247521-100
ABCAAB247521-1EA 40660 PLN
ABCAAB247521-1 ABCAAB247521-100
Anti-Vitamin D Binding protein Rabbit Monoclonal Antibody [clone: EP2923Y]
Przeciwciała
Rabbit monoclonal [EP2923Y] to Vitamin D Binding protein - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Vitamin D Binding protein
Clonality: Monoclonal
Clone: EP2923Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR