Drukuj

Anti-SARS-CoV-2 nucleocapsid protein Rabbit Monoclonal Antibody [clone: EPR24334-118] (Alexa Fluor® 647)

Dostawca: Abcam
AB302552-100 AB302552-10
ABCAAB302552-100EA 3560 PLN
ABCAAB302552-100 ABCAAB302552-10
Anti-SARS-CoV-2 nucleocapsid protein Rabbit Monoclonal Antibody [clone: EPR24334-118] (Alexa Fluor® 647)
Przeciwciała
Alexa Fluor® 647 Rabbit monoclonal [EPR24334-118] to SARS-CoV-2 nucleocapsid protein.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS-CoV-2 nucleocapsid protein
Clonality: Monoclonal
Clone: EPR24334-118
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR