Drukuj

Anti-eIF4B (phospho S504) Rabbit Monoclonal Antibody [clone: EP2261Y]

Dostawca: Abcam
AB284720-100 AB284720-1
ABCAAB284720-100EA 4440 PLN
ABCAAB284720-100 ABCAAB284720-1
Anti-eIF4B (phospho S504) Rabbit Monoclonal Antibody [clone: EP2261Y]
Przeciwciała
Rabbit monoclonal [EP2261Y] to eIF4B (phospho S504) - BSA and Azide free.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: eIF4B
Clonality: Monoclonal
Clone: EP2261Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR