Drukuj

Anti-Cardiac Troponin I Rabbit Monoclonal Antibody [clone: EPR20307] (Alexa Fluor® 488)

Dostawca: Abcam
AB309161-100UL AB309161-10UL
ABCAAB309161-100ULEA 3640 PLN
ABCAAB309161-100UL ABCAAB309161-10UL
Anti-Cardiac Troponin I Rabbit Monoclonal Antibody [clone: EPR20307] (Alexa Fluor® 488)
Przeciwciała
Alexa Fluor® 488 Rabbit monoclonal [EPR20307] to Cardiac Troponin I.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cardiac Troponin I
Clonality: Monoclonal
Clone: EPR20307
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR