Drukuj
Kolumny kapilarne, OPTIMA® 1-TG / OPTIMA® 17-TG
Dostawca: MACHEREY-NAGEL
726131.10 726131.25 726132.10 726132.25 726133.10 726133.25
MANA726131.10EA 1390 PLN
MANA726131.10 MANA726131.25 MANA726132.10 MANA726132.25 MANA726133.10 MANA726133.25
Kolumny kapilarne, OPTIMA® 1-TG / OPTIMA® 17-TG
Kolumny chromatograficzne
Special GC phases for the analysis of triglycerides.
OPTIMA® 1-TG: 100% dimethylpolysiloxane. Offers separation according to carbon number.
USP code: G1 / G2 / G38.
OPTIMA® 17-TG: 50% phenyl - 50% methylpolysiloxane. Offers separation according to degree of unsaturation.
USP code: G3.
Max. temperature for both phases: 370 °C